Learn More
TEAD2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21159-1-AP-150UL
This item is not returnable.
View return policy
Description
TEAD2 is a putative transcription factor that binds to the SPH and GT-IIC enhansons (5′-GTGGAATGT-3′). TEAD2 may be involved in the gene regulation of neural development. TEAD2 binds to the M-CAT motif.Specifications
| TEAD2 | |
| Polyclonal | |
| Unconjugated | |
| TEAD2 | |
| ETF, TEA domain family member 2, TEAD 2, TEAD2, TEF 4, TEF4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 21677, 8463 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P48301, Q15562 | |
| TEAD2 | |
| TEAD2 Fusion Protein Ag14074 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |