Includes
Four gel cassettes; eight foam sponges, 3mm thick; four foam sponges, 6mm thick; and 25 sheets of blotter paper
- Transfers four mini-gels up to 9 × 10cm in size in about 1L of buffer
- Built-in alumina-covered heat exchanger dissipates heat generated by tank electrotransfer; typical temperature increase: ≤5°C
- 80cm of wire in seven parallel strands on each electrode panel delivers a uniform electrical field
- Hinged, color-coded cassettes allow quick setup of transfers
- Replacement electrode panel, sponges, and gel cassettes available separately
Specifications
Specifications
| Certifications/Compliance | EN61010-1, CE 73/23/EEC (LV directive) |
| Type | TE 22 Mini Tank Transfer Unit |
| Includes | Four gel cassettes, eight 3mm thick Foam sponges, four 6mm thick Foam sponges and 25 sheets of blotter Paper, Transphor tank |
| No. of Gels | Transfers four mini-gels up to 9 x 10cm |
| Volume (English) Buffer | 33.81 oz. |
| Volume (Metric) Buffer | 1000 mL |
| Width (English) | 5.5 in. |
| Width (Metric) | 14 cm |
| Depth (English) | 6.5 in. |
| Depth (Metric) | 16.5 cm |
| Show More |