Learn More
TDP-43 (C-terminal) Mouse anti-Human, Unconjugated, Clone: 1G5F6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67345-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
The TDP-43 (C-terminal) antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human TDP-43 (C-terminal). This antibody recognizes human, mouse, rat antigen. The TDP-43 (C-terminal) antibody has been validated for the following applications: FC, IF, IHC, WB, ELISA analysis.
Specifications
TDP-43 (C-terminal) | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry | |
1G5F6 | |
Unconjugated | |
150 μL | |
23435 | |
IgG2a |