Learn More
TDP-43 (C-terminal) Mouse anti-Human, Unconjugated, Clone: 1G5F6, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67345-1-IG-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16889925
  • 1611.84 NOK / 20µL
Estimated Shipment: 13-08-2024
to see stock.
Description
Description
The TDP-43 (C-terminal) antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human TDP-43 (C-terminal). This antibody recognizes human, mouse, rat antigen. The TDP-43 (C-terminal) antibody has been validated for the following applications: FC, IF, IHC, WB, ELISA analysis.
Specifications
Specifications
TDP-43 (C-terminal)
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry
1G5F6
Unconjugated
20 μL
23435
IgG2a