Learn More
TD-198946, MedChemExpress
Product Code. 16492787
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
Product Code. 16492787
Brand: Medchem Express HY15642
MedChemExpress TD-198946, a thienoindazole derivative, is a potent chondrogenic agent.
TD-198946, a thienoindazole derivative, is a potent chondrogenic agent.
Specifications
Chemical Name or Material TD-198946
CAS 364762-86-7
Color Brown
Purity Grade Notes Research
Molecular Formula C27H22N4O3S
Quantity 50 mg
Solubility Information DMSO : ≥ 28 mg/mL (58.03 mM)
SMILES O=C(C1=C2CCC3=C(N(C)N=C3)C2=C(OC4=CC=C(OCC5=NC6=CC=CC=C6C=C5)C=C4)S1)N
Molecular Weight (g/mol) 482.55
Formula Weight 482.55
Percent Purity 98.34%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.