missing translation for 'onlineSavingsMsg'
Learn More

TCV-309 chloride, MedChemExpress

Product Code. 16407586
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
Product Code. Quantity unitSize
16407586 100 mg 100mg
16493687 50 mg 50mg
16491769 1 mg 1mg
16480158 5 mg 5mg
16467698 10 mg 10mg
5 options
This item is not returnable. View return policy

Product Code. 16407586

Brand: Medchem Express HY19121A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TCV-309 chloride is a potent and specific platelet activating factor (PAF) antagonist. TCV-309 chloride specifically inhibits PAF-induced aggregation of rabbit and human platelets, and [3H]PAF binding to rabbit platelet microsomes with IC50 values of 33 nM, 58 nM and 27 nM, respectively. TCV-309 chloride has beneficial effects in anaphylactic shock.

TCV-309 chloride is a potent and specific platelet activating factor (PAF) antagonist. TCV-309 chloride specifically inhibits PAF-induced aggregation of rabbit and human platelets, and [3H]PAF binding to rabbit platelet microsomes with IC50 values of 33 nM, 58 nM and 27 nM, respectively. TCV-309 chloride has beneficial effects in anaphylactic shock.

Specifications

Chemical Name or Material TCV-309 chloride
CAS 121494-09-5
Color Yellow
Purity Grade Notes Research
Molecular Formula C30H34BrClN4O4
Quantity 100 mg
Solubility Information DMSO : ≥ 6.3 mg/mL (10.00 mM)
SMILES CCC[N+]1=CC(C(N(CCC(NCCOC(N2CC3=C(C=CC=C3)CC2)=O)=O)C4=CC=CC=C4)=O)=CC(Br)=C1.[Cl-]
Molecular Weight (g/mol) 629.97
Formula Weight 629.97
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.