missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ TCN2 Recombinant Protein
Brand: Abnova™ H00006948-P01.25ug
Additional Details : Weight : 0.00010kg
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
AAH01176 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
70.62 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
MCEIPEMDSHLVEKLGQHLLPWMDRLSLEHLNPSIYVGLRLSSLQAGTKEDLYLHSLKLGYQQCLLGSAFSEDDGDCQGKPSMGQLALYLLALRANCEFVRGHKGDRLVSQLKWFLEDEKRAIGHDHKGHPHTSYYQYGLGILALCLHQKRVHDSVVDKLLYAVEPFHQGHHSVDTAAMAGLAFTCLKRSNFNPGRRQRITMAIRTVQEEILKAQTPEGHFGNVYSTPLALQFLMTSPMRGAELGTACLKARVALLASLQDGAFQNALMISQLLPVLNHKTYIDLIFPDCLAPRVMLEPAAETIPQTQEIISVTLQVLSLLPPYRQSISVLAGSTVEDVLKKAHELGGFTYETQASLSGPYLTSVMGKAAGEREFWQLLRDPNTPLLQGIADYRPKDGETIELRLVSW | |
D22S676/D22S750/TC2 | |
TCN2 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
6948 | |
TCN2 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
TCN2 | |
Human | |
Recombinant | |
Solution |