missing translation for 'onlineSavingsMsg'
Learn More

TCEAL7 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 11218-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16898143

  • 3828.53 NOK / 150µL
Estimated Shipment: 10-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Plays a role in the negative regulation of NF-kappa-B signaling at the basal level by modulating transcriptional activity of NF-kappa-B on its target gene promoters. Associates with cyclin D1 promoter containing Myc E-box sequence and transcriptionally represses cyclin D1 expression. Regulates telomerase reverse transcriptase expression and telomerase activity in both ALT (alternative lengthening of telomeres)and telomerase-positive cell lines.
Specifications

Specifications

TCEAL7
Polyclonal
Unconjugated
TCEAL7
MPMGp800C04260Q003, TCEA like protein 7, TCEAL7
Rabbit
Antigen Affinity Chromatography
RUO
56849
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9BRU2
TCEAL7
TCEAL7 Fusion Protein Ag1715
150 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers