Learn More
Tocris Bioscience™ TC NTR1 17
GREENER_CHOICE
Selective non-peptide NTS1 agonist
Brand:  Tocris Bioscience™ 5087/50
Encompass
Product Code. 15716456
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
TC NTR1 17 is a non-peptide neurotensin receptor 1 (NTS1) partial agonist (EC50 <15.6 nM and Emax = 63% of neurotensin response in Ca2+ mobilization assay). Exhibits over 50-fold selectivity for NTS1 over NTS2 and GPR35.
Specifications
Specifications
>99%
522.98
50 mg
TC NTR1 17
TC NTR1 17