Learn More
Tocris Bioscience™ TC NTR1 17
Selective non-peptide NTS1 agonist
Brand: Tocris Bioscience™ 5087/50
This item is not returnable.
View return policy
Description
TC NTR1 17 is a non-peptide neurotensin receptor 1 (NTS1) partial agonist (EC50 <15.6 nM and Emax = 63% of neurotensin response in Ca2+ mobilization assay). Exhibits over 50-fold selectivity for NTS1 over NTS2 and GPR35.Specifications
| >99% | |
| 522.98 | |
| 50 mg |
| TC NTR1 17 | |
| TC NTR1 17 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only