Learn More
Thermo Scientific Chemicals TBE running buffer (10X) Product Code.: 15475089
Thermo Scientific Chemicals TBE running buffer (10X)
Product Code. 15475089
4 L, 4 L
Click to view available options
Quantity:
1 L
2 L
4 L
Unit Size:
1 L
2 L
4 L
Product Code. 15475089
Brand: Thermo Scientific Chemicals J62788.K7
Request Bulk or Custom Format
Commonly used in nucleic acid electrophoresis. TBE Buffer is used for polyacrylamide and agarose gel electrophoresis. It is ideally suited for DNA and RNA separation on longer run, higher voltage or amperage gels. TBE is used with non-denaturing or denaturing (7 M urea) gels.
TBE (Tris/Borate/EDTA) Buffer is commonly used in nucleic acid electrophoresis. TBE Buffer is used for polyacrylamide and agarose gel electrophoresis. It is ideally suited for DNA and RNA separation on longer run, higher voltage or amperage gels. TBE is used with non-denaturing or denaturing (7 M urea) gels. It is also routinely used for DNA automated sequencing gel.5 TBE can also be used for agarose gels but is not recommended for preparative gels for recovery of nucleic acids. Tris-Borate-EDTA buffer has been used for pulsed-field gel electrophoresis (PFGE).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material TBE running buffer (10X)
Physical Form Liquid
Quantity 4 L
Packaging Plastic bottle
Solubility Information It is soluble in water.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot NumberU07L519 Certificate TypeCertificate of Analysis Date29/07/2025 Product CodeJ62788.K7,J62788.K2,J62788.K3
Lot NumberP29L528 Certificate TypeCertificate of Analysis Date17/05/2025 Product CodeJ62788.K7,J62788.K2,J62788.K3
Lot NumberQ20L504 Certificate TypeCertificate of Analysis Date17/05/2025 Product CodeJ62788.K7,J62788.K2,J62788.K3
Lot NumberR24L529 Certificate TypeCertificate of Analysis Date17/05/2025 Product CodeJ62788.K7,J62788.K2,J62788.K3
Lot NumberN25L510 Certificate TypeCertificate of Analysis Date19/02/2025 Product CodeJ62788.K7,J62788.K2,J62788.K3
compliance-icons compliance-icons
Product Identifier
  • TBE running buffer (10X)
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : N,N-1,2-ethanediylbis[N-(carboxymethyl)-, disodium salt, dihydrate
Product Title
Thermo Scientific Chemicals TBE running buffer (10X) >