missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBCE Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12021-1-AP-20UL
This item is not returnable.
View return policy
Description
Cofactor E is one of four proteins (cofactors A, D, E, and C) involved in the pathway leading to correctly folded beta-tubulin from folding intermediates. Cofactors A and D are believed to play a role in capturing and stabilizing beta-tubulin intermediates in a quasi-native confirmation. Cofactor E binds to the cofactor D/beta-tubulin complex; interaction with cofactor C then causes the release of beta-tubulin polypeptides that are committed to the native state.Specifications
| TBCE | |
| Polyclonal | |
| Unconjugated | |
| TBCE | |
| HRD, KCS, KCS1, pac2, TBCE, tubulin folding cofactor E, Tubulin specific chaperone E | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 361255, 6905, 70430 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q15813, Q5FVQ9, Q8CIV8 | |
| Tbce | |
| TBCE Fusion Protein Ag2650 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction