missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBCB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15782-1-AP-150UL
This item is not returnable.
View return policy
Description
CKAP1 binds to alpha-tubulin folding intermediates after their interaction with cytosolic chaperonin in the pathway leading from newly synthesized tubulin to properly folded heterodimer. Involved in regulation of tubulin heterodimer dissociation. May function as a negative regulator of axonal growth.Specifications
| TBCB | |
| Polyclonal | |
| Unconjugated | |
| TBCB | |
| CG22, CKAP1, CKAPI, TBCB, tubulin folding cofactor B, Tubulin specific chaperone B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1155, 292777, 66411 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q99426, Q9D1E6 | |
| Tbcb | |
| TBCB Fusion Protein Ag8473 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction