missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBC1D15 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17252-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Acts as a regulator of transcriptional activation. Plays a role in the inducible expression of cytokine genes in T-cells, especially in the induction of the IL-2.Specifications
TBC1D15 | |
Polyclonal | |
Unconjugated | |
TBC1D15 | |
GAP for RAB7, GTPase activating protein RAB7, Rab7 GAP, TBC1 domain family member 15, TBC1 domain family, member 15, TBC1D15 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
366896, 64786, 66687 | |
-20°C | |
Liquid |
Western Blot | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8TC07, Q9CXF4 | |
Tbc1d15 | |
TBC1D15 Fusion Protein Ag10625 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |