Learn More
TBA-7371, MedChemExpress
Product Code. 16487696
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16487696
Brand: Medchem Express HY19750
MedChemExpress TBA-7371 is a potent, noncovalent DprE1 inhibitor. TBA-7371 has potent antitubercular activity .
TBA-7371 is a potent, noncovalent DprE1 inhibitor. TBA-7371 has potent antitubercular activity .
Specifications
Chemical Name or Material TBA-7371
CAS 1494675-86-3
Color White
Purity Grade Notes Research
Molecular Formula C18H21N5O3
Quantity 100 mg
Solubility Information DMSO : 25 mg/mL (70.35 mM; Need ultrasonic)
SMILES O=C(C1=CN(CC2=NC=NC(OC)=C2C)C3=CC(C)=CN=C31)NCCO
Molecular Weight (g/mol) 355.39
Formula Weight 355.39
Percent Purity 99.64%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.