missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tazarotene, MedChemExpress
Description
Tazarotene (AGN 190168) is a selective retinoic acid receptor (RAR) agonist for the treatment of plaque psoriasis and acne vulgaris.
Specifications
Specifications
| Chemical Name or Material | Tazarotene |
| Physical Form | Solid |
| CAS | 118292-40-3 |
| Color | Light Yellow |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C21H21NO2S |
| For Use With (Application) | COVID-19-immunoregulation |
| Synonym | AGN 190168 |
| Solubility Information | DMSO : 50 mg/mL (142.26 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?