Learn More
TAX1BP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Product Code. 16858943 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16858943 20 μL 20µL
16848943 150 μL 150µL
2 options
Product Code. 16858943
Brand: Proteintech 116921AP20UL
Rabbit Polyclonal Antibody
May regulate a number of protein-protein interactions by competing for PDZ domain binding sites. Binds CTNNB1 and may thereby act as an inhibitor of the Wnt signaling pathway. Competes with LIN7A for KCNJ4 binding, and thereby promotes KCNJ4 internalization. May play a role in the Rho signaling pathway. May play a role in activation of CDC42 by the viral protein HPV16 E6.
Specifications
Antigen TAX1BP3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.47 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene TAX1BP3
Gene Accession No. O14907, Q4QQV1, Q9DBG9
Gene Alias Tax interaction protein 1, Tax1 binding protein 3, TAX1BP3, TIP 1, TIP1
Gene Symbols TAX1BP3
Host Species Rabbit
Immunogen TAX1BP3 Fusion Protein Ag2306
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30851, 360564, 76281
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG