Learn More
Taurinamide Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Taurinamide Hydrochloride |
| CAS | 89756-60-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C2 H8 N2 O2 S . HCl |
| Quantity | 1 g |
| Synonym | 2-Amino-ethanesulfonamide Hydrochloride, 2-Aminoethanesulfonamide Hydrochloride, 2-Sulfamoylethylamine Hydrochloride, Aminoethanesulfonamide Hydrochloride, NSC 403678 Hydrochloride |
| InChI Formula | InChI=1S/C2H8N2O2S.ClH/c3-1-2-7(4,5)6;/h1-3H2,(H2,4,5,6);1H |
| SMILES | NCCS(N)(=O)=O.Cl |
| IUPAC Name | 2-aminoethanesulfonamide;hydrochloride |
| Molecular Weight (g/mol) | 160.62 |
| Show More |