Learn More
Tartrate solution for Ion Chromatography 1g/L H2O matrix, Fisher chemical™
BIOPHARMA
Product Code. 12982205 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Packaging:
Bottle
Unit Size:
100mL
250mL
500mL
Product Code. Quantity Packaging unitSize
12982205 100 mL Bottle 100mL
12992205 250 mL Bottle 250mL
12902215 500 mL Bottle 500mL
3 options
Product Code. 12982205
Brand: Fisher Chemical A 047 100
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Chemical Identifiers
Molecular Formula OOC(CHOH)2COO2-
Specifications
Packaging Bottle
Physical Form Liquid
Concentration 1 g/L
Type Single-component Standard Solution
Solvent or Matrix Water
Quantity 100 mL
Chemical Name or Material Tartrate