Learn More
Buerkle™ Target Immersion Cylinder
Shop All Bürkle Products
Click to view available options
Capacity (Metric):
500 mL
Unit Size:
Each
Description
The Target immersion cylinder is uses for precise liquid sampling.
- Weight 2. 5 kg
- Includes venting lowering cable (5m), valve and cable reel
- Screw-off top enables easy cleaning
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Diameter (Metric) Outer | 75 mm |
| Height (Metric) | 210 mm |