Learn More
MP Biomedicals™ Taq Core Kit 25
Product Code. 11477030 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
3 x 250 u
Unit Size:
750 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11477030 3 x 250 u 750 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11477030 Supplier MP Biomedicals™ Supplier No. 11EPTQK253
  • ONLINE EXCLUSIVE PRICE
    7675.00 NOK
    7307.23 NOK / Pack of 750
    Save 367.77 NOK
    4% Off
 Restricted Item
Estimated Shipment: 09-11-2026
Taq DNA Polymerase + dNTPs in one kit
  • Taq CORE Kits contain all the reagents (presented as separate items) required for PCR:
  • MP Biomedicals high quality recombinant Taq DNA polymerase, high purity dNTPs Mix (>99%), optimized incubation buffers with and without MgCl2 and a separate MgCl2 solution.
  • dNTPs at 25mM each
Specifications
Quantity 3 x 250 u