Learn More
Sonics Materials™ Tapered Microtip
Click to view available options
Diameter (Metric):
5 mm
6 mm
Unit Size:
Each
Specifications
Specifications
| Amplitude | 180 μm |
| Product Type | Tapered Microtip |
| Diameter (Metric) | 6 mm |
| Packaging Quantity | 1 Ea. |