missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAOK3/KDS Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | TAOK3/KDS |
|---|---|
| Concentration | 600 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17689626
|
Proteintech
28403-1-AP-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17679626
|
Proteintech
28403-1-AP-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TAOK3/KDS Polyclonal antibody apecifically detetects TAOK3/KDS antigen in Human Samples. It is validated for Western Blot, ELISA.Specifications
| TAOK3/KDS | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| 51347 | |
| TAOK3 fusion protein Ag29158 | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 600 μg/mL | |
| Polyclonal | |
| Liquid | |
| Human | |
| BC002756 | |
| TAOK3 | |
| IgG | |
| Antigen affinity purification |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title