missing translation for 'onlineSavingsMsg'
Learn More

TAOK3/KDS Rabbit anti-Human, Polyclonal, Proteintech

Product Code. 17689626 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
17689626 20 μL 20µL
17679626 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 17689626

Brand: Proteintech 284031AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TAOK3/KDS Polyclonal antibody apecifically detetects TAOK3/KDS antigen in Human Samples. It is validated for Western Blot, ELISA.

Specifications

Antigen TAOK3/KDS
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 600 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
Gene Accession No. BC002756
Gene Symbols TAOK3
Host Species Rabbit
Immunogen TAOK3 fusion protein Ag29158
Purification Method Antigen affinity purification
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 51347
Target Species Human
Content And Storage Store at -20°C. Stable for one year after shipment.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.