Learn More
Tanshinone IIA sulfonate sodium, MedChemExpress
Product Code. 16453916
Encompass_Preferred
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
Product Code. 16453916
Brand: Medchem Express HYN1370
MedChemExpress Tanshinone IIA sulfonate (sodium) is a derivative of tanshinone IIA, which acts as an inhibitor of store-operated Ca2+ entry (SOCE), and is used to treat cardiovascular disorders.
Tanshinone IIA sulfonate (sodium) is a derivative of tanshinone IIA, which acts as an inhibitor of store-operated Ca2+ entry (SOCE), and is used to treat cardiovascular disorders.
Specifications
Chemical Name or Material Tanshinone IIA sulfonate sodium
CAS 69659-80-9
Color wine
Purity Grade Notes Research
Molecular Formula C19H17NaO6S
Quantity 25 mg
Synonym Sodium Tanshinone IIA sulfonate Tanshinone IIA sodium sulfonate
Solubility Information DMSO : 62.5 mg/mL (157.67 mM; Need ultrasonic) ∣H2O : 8.33 mg/mL (21.01 mM; Need ultrasonic)
SMILES O=S(C1=C(C)C(C(C(C2=C3C=CC4=C2CCCC4(C)C)=O)=O)=C3O1)(O[Na])=O
Molecular Weight (g/mol) 396.39
Formula Weight 396.39
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.