Learn More
3M™ Tanker Spill Response Kit
Description
- Designed to absorb and contain small spills and leaks
- Compact size is convenient for trucks, shipping departments, and areas of oil transfer
- Kit is easily stowed in confined areas, such as behind the drivers seat
Specifications
Specifications
| Capacity (Metric) | 30 L |
| Type | Sorbent Spill Kit |
| For Use With (Application) | Absorbing/Containing Small Spills and Leaks |
| Includes | (50) Sheets, 2 Minibooms, 1 disposal bag and Tie, (1) Disposal Guide and Label |
| Model | TSK-30 |
| Packaging Quantity | 2/Pk. |
| Dimensions (L x W x H) | 510 x 430 x 210 mm |
| Length (Metric) | 510 mm |
| Width (Metric) | 430 mm |
| Height (Metric) | 210 mm |