Learn More
Invitrogen™ Tango™ OPRM1-bla U2OS Cells
Click to view available options
Quantity:
2 x 106 Cells
Unit Size:
Each
Description
The Tango™ OPRM1-bla U2OS cells contain the human Opioid Receptor Mu 1 (OPRM1) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line. This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase (bla) reporter gene under the control of a UAS response element.
The Tango™ OPRM1-bla U2OS cells have been functionally validated for Z' factor and EC50 concentrations of DAMGO. In addition, Tango™ OPRM1-bla U2OS cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary in the Manuals and Brochures section.
The Tango™ OPRM1-bla U2OS cells have been functionally validated for Z' factor and EC50 concentrations of DAMGO. In addition, Tango™ OPRM1-bla U2OS cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary in the Manuals and Brochures section.
Specifications
Specifications
| Accession Number | NM_000914 |
| Assay Entry | Cell-based beta-arrestin recruitment |
| Cell State | Dividing Cells |
| Content And Storage | (LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol). Includes: o Tango™ OPRM1-bla U2OS cells o Tango™ OPRM1-bla U2OS Assay Protocol o Certificate of Analysis The Tango™ OPRM1-bla U2OS cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt. |
| Cytoplasmic Tail | Tail from AVPR2 GPCR |
| Detection Method | Fluorescence |
| Gene Symbol | OPRM1 |
| Population Status | Single Cell Clone Origin |
| Product Line | Tango |
| Quantity | 2 x 106 Cells |
| Show More |