Learn More
Thermo Scientific™ Tango™ CXCR4-bla U2OS Cells
Product Code. 10379093
Click to view available options
Quantity:
8 x 106 Cells
Unit Size:
Each
Product Code. 10379093
Brand: Thermo Scientific™ K1779
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The Tango™ CXCR4-bla U2OS cells contain the human Chemokine (C-X-C motif) receptor 4 (CXCR4) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line.
The Tango™ CXCR4-bla U2OS cells contain the human Chemokine (C-X-C motif) receptor 4 (CXCR4) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line. This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase (bla) reporter gene under the control of a UAS response element.
The Tango™ CXCR4-bla U2OS cells are functionally validated for Z' and EC50 concentrations of SDF1α. In addition, Tango™ CXCR4-bla U2OS cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.
For Academic pricing please login or contact us at discoverysciences@invitrogen.com.
TRUSTED_SUSTAINABILITY
Specifications
Accession Number NM_003467.2
Assay Entry Cell-based beta-arrestin recruitment
Cell State Dividing Cells
Content And Storage LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.
Includes:
• Tango™ CXCR4-bla U2OS cells
• Tango™ CXCR4-bla U2OS Assay Protocol
• Certificate of Analysis
The Tango™ CXCR4-bla U2OS cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Cytoplasmic Tail Native Tail
Description Tango™ CXCR4-bla U2OS Cells, 8 x 106 Cells, ∽8,000,000 cells/vial at 8,000,000 cells/mL Quantity of Cells, U2OS Cell Line, Fluorescent Detection Method, GPCRs Druggable Target, BLA (Beta-Lactamase) Reporter, Liquid Nitrogen Storage
Detection Method Fluorescence
For Use With (Application) FRET
Gene Symbol CXCR4
Includes Tango™ CXCR4-bla U2OS cells (LiveBLAzer™-FRET B/G loading kit, solution D and other materials are required separately; please refer to the protocol). Includes: Tango™ CXCR4-bla U2OS cells, Tango™ CXCR4-bla U2OS assay protocol, certificate of analysis
Population Status Single Cell Clone Origin
Product Line GeneBLAzer, Tango
Quantity 8 x 106 Cells
Readout End Point
Reporter Gene BLA (Beta-Lactamase)
Sample Type U2OS
Target Entry CXCR4
Type U2OS Cell
Cell Line U2OS
System Type Tango™
Druggable Target GPCR