Learn More
Thermo Scientific™ Tango™ CMKLR1-bla U2OS Cells
Product Code. 10701434
Change view
Click to view available options
Quantity:
2 x 106 Cells
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10701434 2 x 106 Cells Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10701434 Supplier Thermo Scientific™ Supplier No. K1527
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The Tango™ CMKLR1-bla U2OS cells contain the human Chemokine like receptor 1 (CMKLR1) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line.
The Tango™ CMKLR1-bla U2OS cells contain the human Chemokine like receptor 1 (CMKLR1) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line. This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase (bla) reporter gene under the control of a UAS response element.
TRUSTED_SUSTAINABILITY
Specifications
Accession Number NM_004072.1
Assay Entry Cell-based beta-arrestin recruitment
Cell State Dividing Cells
Content And Storage LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.
Includes:
• Tango™ CMKLR1-bla U2OS cells
• Tango™ CMKLR1-bla U2OS Assay Protocol
• Certificate of Analysis
The Tango™ CMKLR1-bla U2OS cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Cytoplasmic Tail Native Tail
Description Tango™ CMKLR1-bla U2OS Cells, 2 x 106 Cells, U2OS Cell Line, Fluorescent Detection Method, GPCRs Druggable Target, BLA (Beta-Lactamase) Reporter, Liquid Nitrogen Storage
Detection Method Fluorescence
For Use With (Application) Pharma and biopharma, drug discovery and development, target and lead identification and validation, G-protein coupled receptor biology, cell-based GPCR reporter assays
Gene Symbol CMKLR1
Includes Tango™ CMKLR1-bla U2OS cells (LiveBLAzer™-FRET B/G loading kit, solution D and other materials are required separately; please refer to the protocol). Includes: Tango™ CMKLR1-bla U2OS cells, Tango™ CMKLR1-bla U2OS assay protocol, certificate of analysis
Population Status Single Cell Clone Origin
Product Line Tango
Quantity 2 x 106 Cells
Reporter Gene BLA (Beta-Lactamase)
Sample Type U2OS
Shipping Condition Room Temperature
Target Entry CMKLR1
Type U2OS Cell
Cell Line U2OS
System Type Tango™
Druggable Target GPCR