Learn More
Tanaproget, MedChemExpress
Description
Tanaproget (NSP-989) is a novel nonsteroidal progesterone receptor agonist which can bind to the PR from various species with a higher relative affinity than reference steroidal progestins.
Specifications
Specifications
| Chemical Name or Material | Tanaproget |
| CAS | 304853-42-7 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C16H15N3OS |
| Quantity | 50 mg |
| Synonym | NSP-989 |
| Solubility Information | DMSO : ≥ 50 mg/mL (168.14 mM) |
| SMILES | N#CC1=CC=C(C2=CC=C(NC(OC3(C)C)=S)C3=C2)N1C |
| Molecular Weight (g/mol) | 297.37 |
| Show More |