Learn More
- Home
- Products
- Specialty Lab Equipment, Instruments, and Apparatuses
- Crucibles
- Thermo Scientific Chemicals Al-23 Crucible, Tammann;Height x OD x ID (mm), 100 x 12 x 8;Volume (ml), 5;Tammann
Thermo Scientific Chemicals Al-23 Crucible, Tammann;Height x OD x ID (mm), 100 x 12 x 8;Volume (ml), 5;Tammann
Product Code. 15433555| Product Code. | Capacity (Metric) | Diameter (Metric) Outer | Diameter (Metric) Inner | Height (Metric) | unitSize |
|---|---|---|---|---|---|
| 15433555 | 490 mL | 47mm | 280mm | 55mm | Each |
| 15403545 | 0.2 mL | 3mm | 30mm | 6mm | Each |
| 15423545 | 2.3 mL | 6mm | 80mm | 10mm | Each |
| 15433545 | 5 mL | 8mm | 100mm | 12mm | Each |
| 15443545 | 8 mL | 10mm | 100mm | 15mm | Each |
| 15413545 | 1 mL | 5mm | 50mm | 8mm | Each |
| 15463545 | 17 mL | 15mm | 100mm | 20mm | Each |
| 15483545 | 90 mL | 20mm | 280mm | 24mm | Each |
| 15493545 | 30 mL | 20mm | 100mm | 25mm | Each |
| 15443555 | 790 mL | 58mm | 280mm | 68mm | Each |
| 15403555 | 123 mL | 28mm | 190mm | 34mm | Each |
| 15423555 | 300 mL | 38mm | 280mm | 44mm | Each |
| 15413555 | 180 mL | 28mm | 280mm | 34mm | Each |
| 15453555 | 1070 mL | 58mm | 390mm | 68mm | Each |
Product Code. 15433555
Brand: Thermo Scientific Chemicals 033065.KF
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Diameter (Metric) Inner | 280mm |
| Material | Al-23 |
| Quantity | 1 Piece |
| Capacity (Metric) | 490 mL |
| Diameter (Metric) Outer | 47mm |
| Height (Metric) | 55mm |
| Product Type | Tammann Al-23 Crucibles |