Learn More
Thermo Scientific Chemicals Al-23 Crucible, Tammann;Height x OD x ID (mm), 100 x 12 x 8;Volume (ml), 5;Tammann
Product Code. 15413555
Change view
Click to view available options
Capacity (Metric):
0.2 mL
1 mL
2.3 mL
5 mL
8 mL
17 mL
30 mL
90 mL
123 mL
180 mL
300 mL
490 mL
790 mL
1070 mL
Diameter (Metric) Outer:
3mm
5mm
6mm
8mm
10mm
15mm
20mm
28mm
38mm
47mm
58mm
Diameter (Metric) Inner:
30mm
50mm
80mm
100mm
190mm
280mm
390mm
Height (Metric):
6mm
8mm
10mm
12mm
15mm
20mm
24mm
25mm
34mm
44mm
55mm
68mm
Unit Size:
Each
Product Code. Capacity (Metric) Diameter (Metric) Outer Diameter (Metric) Inner Height (Metric) unitSize
15413555 180 mL 28mm 280mm 34mm Each
15403545 0.2 mL 3mm 30mm 6mm Each
15423545 2.3 mL 6mm 80mm 10mm Each
15433545 5 mL 8mm 100mm 12mm Each
15443545 8 mL 10mm 100mm 15mm Each
15413545 1 mL 5mm 50mm 8mm Each
15463545 17 mL 15mm 100mm 20mm Each
15483545 90 mL 20mm 280mm 24mm Each
15493545 30 mL 20mm 100mm 25mm Each
15443555 790 mL 58mm 280mm 68mm Each
15403555 123 mL 28mm 190mm 34mm Each
15423555 300 mL 38mm 280mm 44mm Each
15433555 490 mL 47mm 280mm 55mm Each
15453555 1070 mL 58mm 390mm 68mm Each
14 options
Product Code. 15413555
Brand: Thermo Scientific Chemicals 033063.KF
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Inner 280mm
Material Al-23
Quantity 1 Piece
Capacity (Metric) 180 mL
Diameter (Metric) Outer 28mm
Height (Metric) 34mm
Product Type Tammann Al-23 Crucibles