Learn More
Thermo Scientific Chemicals Al-23 Crucible, Tammann;Height x OD x ID (mm), 100 x 12 x 8;Volume (ml), 5;Tammann
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Diameter (Metric) Inner | 50mm |
| Material | Al-23 |
| Quantity | 1 Piece |
| Capacity (Metric) | 1 mL |
| Diameter (Metric) Outer | 5mm |
| Height (Metric) | 8mm |
| Product Type | Tammann Al-23 Crucibles |
Frequently Asked Questions (FAQs)
The maximum working temperature for the Al-23 Tube is 1950 degrees C.