Learn More
Köttermann Tall Safety Storage Cabinet with Drawers
Product Code. p-7236286
Click to view available options
No. of Doors:
1
2
No. of Shelves:
4
5
8
9
10
Unit Size:
Each
Product Code. 17322139
Brand: Köttermann 37100036
This item requires a price update prior to being ordered.
Request a Price for this item here.
This Tall Safety Storage Cabinet offers top safety for flammable hazardous substances, complying with fire resistance class standards (Type 90).
  • Different versions that meet your needs
  • Additional storage space
  • Top safety for flammable hazardous substances (Type 90)
Specifications
Dimensions (D x W x H) 853 x 450 x 1966 mm
Door Style Drawer
Includes 1 Drip Tray
No. of Doors 1
No. of Shelves 4