Learn More
Köttermann Tall Safety Storage Cabinet with Drawers
Description
- Different versions that meet your needs
- Additional storage space
- Top safety for flammable hazardous substances (Type 90)
Specifications
Specifications
| Dimensions (D x W x H) | 853 x 815 x 1966 mm |
| Door Style | Drawer |
| Includes | 1 Drip Tray |
| No. of Doors | 2 |
| No. of Shelves | 10 |