Learn More
Kartell™ Tall Form Graduated Measuring Cylinders
Click to view available options
Graduations:
2 mL
5 mL
10 mL
20 mL
100 mL
200 mL
Unit Size:
Pack of 10
Pack of 12
Pack of 2
Pack of 30
Pack of 6
Capacity (Metric):
10 mL
50 mL
Description
- Tall form
- Circular base
- Good chemical resistance
- Permanent graduations moulded into cylinders
- Can withstand temperatures up to 170°C
- Suitable for applications with food
- No wetting and no meniscus
Specifications
Specifications
| Class | B |
| Graduations | 10 mL |
| Autoclavable | Autoclavable |
| Max. Temperature | 170°C |
| Subdivision | 1mL |
| Shape | Cylindrical |
| For Use With (Application) | Foodstuff |
| Scale Color | Blue |
| Height (Metric) | 249 mm |
| Type | Graduated Measuring Cylinder |