Learn More
BRAND™ Tall Form Borosilicate Glass Beakers
Shop All Brand Products
Click to view available options
Capacity (Metric):
50 mL
100 mL
150 mL
250 mL
400 mL
600 mL
1000 mL
Unit Size:
Pack of 10
Description
- With graduation and spout
- Provided in packs of 10
- With write-on label area
- Acc. to DIN 12331/ISO 3819
Specifications
Specifications
| Color | Clear |
| Diameter (Metric) Outer | 80 mm |
| Capacity (Metric) | 600 mL |
| Material | Borosilicate Glass |
| Height (Metric) Outer | 150 mm |
| Quantity | 10/Pk. |
| Type | Beaker |
| Graduated | Yes |
| Scale | Printed Scale |
| Style | Tall Form |