Learn More
Talc, TRC
Specifications
Specifications
| Chemical Name or Material | Talc |
| CAS | 14807-96-6 |
| Molecular Formula | 3 Mg O . 4 O2 Si . H2 O |
| Quantity | 250 g |
| Synonym | Talc, hydrated magnesium silicate |
| InChI Formula | InChI=1S/3Mg.4O2Si.H2O.3O/c;;;4*1-3-2;;;;/h;;;;;;;1H2;;; |
| SMILES | O.O=[Mg].O=[Mg].O=[Mg].O=[Si]=O.O=[Si]=O.O=[Si]=O.O=[Si]=O |
| IUPAC Name | dioxosilane;oxomagnesium;hydrate |
| Molecular Weight (g/mol) | 379.27 |
| Formula Weight | 377.8175 |