Learn More
OHAUS™ Support Rod and Clamp Kit
Shop All Ohaus Products
Click to view available options
Color:
Silver
Unit Size:
Each
Description
- For all Hotplates or Hotplate-Stirrers
- Includes an 45.7cm (18 in.), Stainless-steel support rod, thermometer/temperature probe extension clamp, 3-prong medium swivel clamp and hook connector
Compliance
UL, cUL, CE marked
Specifications
Specifications
| Quantity | Ea. |
| Type | Clamp Kit |
| Material | Nickel-plated |
| Includes | 18 in. support rod, thermometer/temperature probe extension clamp, 3 prong medium swivel clamp and hook connector |
| For Use With (Equipment) | Hotplates, stirrers and hotplate and stirrers |
| Description | Support Rod And Clamp Kit |
| Color | Silver |
| Jaw Type | 3-Prong Medium Swivel Clamp |
| Diameter (Metric) Inside | 0 to 69 mm |