Learn More
(-)-Talarozole, MedChemExpress
Product Code. 16481439
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16481439 1 mg 1mg
16468777 5 mg 5mg
16466178 10 mg 10mg
16432317 50 mg 50mg
16413446 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16481439 Supplier Medchem Express Supplier No. HY14802D
MedChemExpress (-)-Talarozole is a potent inhibitor of retinoic acid metabolism extracted from patent WO 1997049704 A1.
(-)-Talarozole is a potent inhibitor of retinoic acid metabolism extracted from patent WO 1997049704 A1.
Specifications
Chemical Name or Material (-)-Talarozole
Physical Form Solid
CAS 201410-67-5
Color Off-White
Quantity 1 mg
Purity Grade Notes Research
Molecular Formula C21H23N5S
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 72.5 mg/mL (192.05 mM)
SMILES CCC(CC)C(C1=CC=C(NC2=NC3=CC=CC=C3S2)C=C1)N4N=CN=C4.[(-)].[Rotation]
Molecular Weight (g/mol) 377.51
Formula Weight 377.51
Percent Purity 98.02%
Grade Research