missing translation for 'onlineSavingsMsg'
Learn More

TAK1 Rabbit anti-Hamster, Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 12330-2-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16889983

  • 4593.76 NOK / 150µL
Estimated Shipment: 15-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

TAK1 (also known as MAP3K7), a part of the serine/threonine kinase family, is a versatile protein with many signaling functions. Originally identified as a TGF- beta-activated kinase, TAK1 interacts with TGF- beta-receptor and TRAF6 to modulate TGF- beta activation of JNK and p38. It has additional roles in activation of p38 and JNK through Wnt, BMP, and activin signaling pathways as well in response to thyroid hormone, osmotic stress, endothelin, and ephrine. Through activation of von Hippel-Lindau tumor suppressor expression, TAK1 represses PDGF-B, integrin beta1 and integrin beta5, promoting proper wound healing. TAK1 also plays an essential role in IKK activation in numerous signaling pathways including IL-1, IL-6, IL-18, TNF, CD40, TLR, and RIG-I. Activation of the TAK1-IKK pathway requires TAB2/3 and ubiquitination. In this process, TAB2/3 binds to the C-terminal region of TAK1 and becomes polyubiquitinated, TAK1 is autophosphorylated at Thr178, Thr184, Thr187 and Ser192, and finally TAK1 phosphorylates IKK- beta. Additionally, TAK1 represses human telomerase reverse transcriptase suggesting a role in regulation of cell lifespan.
Specifications

Specifications

TAK1
Polyclonal
Unconjugated
MAP3K7
MAP3K7, TAK1, TGF beta activated kinase 1, TGF1a
Rabbit
Antigen Affinity Chromatography
RUO
100774341, 26409, 6885
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O43318, Q62073
MAP3K7
TAK1 Fusion Protein Ag3041
150 μL
Primary
Hamster, Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers