Learn More
Brady™ Lockout Labels
Brand:  Brady™ 256215
Additional Details : Weight : 0.05000kg
 View more versions of this product
Product Code. 11726875
  • 272.00 NOK / 1 pack
Estimated Shipment: 09-08-2024
Specifications
10 Pk.
Black on Yellow
DANGER - DEFENSE D’ENCLENCHER TANT QUE L'ETIQUETTE EST APPLIQUEE
6.3 in.
0.249 mm
-40°F to +212°F
75 mm
7 mm
Tag
Polyester
Lockout Tagout
160 mm
-40°C to +100°C
2.95 in.
160 x 75 mm
Lockout Tagout Tags