Learn More
Tafamidis Meglumine, TRC
Specifications
Specifications
| Chemical Name or Material | Tafamidis Meglumine |
| CAS | 951395-08-7 |
| Molecular Formula | C14H7Cl2NO3 . C7H17NO5 |
| Quantity | 250 mg |
| Synonym | Vyndaqel |
| InChI Formula | InChI=1S/C14H7Cl2NO3.C7H17NO5/c15-9-3-8(4-10(16)6-9)13-17-11-2-1-7(14(18)19)5-12(11)20-13;1-8-2-4(10)6(12)7(13)5(11)3-9/h1-6H,(H,18,19);4-13H,2-3H2,1H3/t;4-,5+,6+,7+/m.0/s1 |
| SMILES | CNC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO.OC(=O)c1ccc2nc(oc2c1)c3cc(Cl)cc(Cl)c3 |
| IUPAC Name | 2-(3,5-dichlorophenyl)-1,3-benzoxazole-6-carboxylic acid;(2R,3R,4R,5S)-6-(methylamino)hexane-1,2,3,4,5-pentol |
| Molecular Weight (g/mol) | 503.3 |
| Formula Weight | 502.091 |