Learn More
Tafamidis Meglumine, TRC
Product Code. 30425296
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity unitSize
30425296 100 mg 100mg
1 options
Product Code. 30425296
Brand: Toronto Research Chemicals TRCT223615100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Tafamidis Meglumine
CAS 951395-08-7
Molecular Formula C14H7Cl2NO3 . C7H17NO5
Quantity 100 mg
Synonym Vyndaqel
InChI Formula InChI=1S/C14H7Cl2NO3.C7H17NO5/c15-9-3-8(4-10(16)6-9)13-17-11-2-1-7(14(18)19)5-12(11)20-13;1-8-2-4(10)6(12)7(13)5(11)3-9/h1-6H,(H,18,19);4-13H,2-3H2,1H3/t;4-,5+,6+,7+/m.0/s1
SMILES CNC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO.OC(=O)c1ccc2nc(oc2c1)c3cc(Cl)cc(Cl)c3
IUPAC Name 2-(3,5-dichlorophenyl)-1,3-benzoxazole-6-carboxylic acid;(2R,3R,4R,5S)-6-(methylamino)hexane-1,2,3,4,5-pentol
Molecular Weight (g/mol) 503.3
Formula Weight 502.091