missing translation for 'onlineSavingsMsg'
Learn More

Tafamidis meglumine, MedChemExpress

Product Code. p-200098124
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16498797

Brand: Medchem Express HY14852A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tafamidis meglumine (Fx-1006A) is a potent and selective transthyretin (TTR) stabilizer, shows comparable potency and efficacy to the mutumant homotetramers V30M-TTR, V122I-TTR and wild type WT-TTR, with EC50s of 2.7-3.2 μM. Tafamidis meglumine inhibits amyloidogenesis.

Tafamidis meglumine (Fx-1006A) is a potent and selective transthyretin (TTR) stabilizer, shows comparable potency and efficacy to the mutumant homotetramers V30M-TTR, V122I-TTR and wild type WT-TTR, with EC50s of 2.7-3.2 μM. Tafamidis meglumine inhibits amyloidogenesis.

Specifications

Chemical Name or Material Tafamidis meglumine
Physical Form Solid
CAS 951395-08-7
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C21H24Cl2N2O8
For Use With (Application) Neuroscience-Neuromodulation
Synonym Fx-1006A
Solubility Information DMSO : 12.5 mg/mL (24.83 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(C1=CC=C2N=C(C3=CC(Cl)=CC(Cl)=C3)OC2=C1)O.O[C@@H]([C@@H]([C@@H](CO)O)O)[C@@H](O)CNC
Molecular Weight (g/mol) 503.33
Formula Weight 503.33
Percent Purity 99.52%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.