Learn More
Tafamidis meglumine, MedChemExpress
Product Code. 16498797
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Packungsgröße:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
16498797 5 mg 5mg
16496566 100 mg 100mg
16472337 50 mg 50mg
16463446 25 mg 25mg
16456208 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 16498797 Lieferant Medchem Express Lieferanten-Nr. HY14852A
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
MedChemExpress Tafamidis meglumine (Fx-1006A) is a potent and selective transthyretin (TTR) stabilizer, shows comparable potency and efficacy to the mutumant homotetramers V30M-TTR, V122I-TTR and wild type WT-TTR, with EC50s of 2.7-3.2 μM. Tafamidis meglumine inhibits amyloidogenesis.
Tafamidis meglumine (Fx-1006A) is a potent and selective transthyretin (TTR) stabilizer, shows comparable potency and efficacy to the mutumant homotetramers V30M-TTR, V122I-TTR and wild type WT-TTR, with EC50s of 2.7-3.2 μM. Tafamidis meglumine inhibits amyloidogenesis.
Spezifikation
Chemical Name or Material Tafamidis meglumine
Physical Form Solid
CAS 951395-08-7
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C21H24Cl2N2O8
For Use With (Application) Neuroscience-Neuromodulation
Synonym Fx-1006A
Solubility Information DMSO : 12.5 mg/mL (24.83 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(C1=CC=C2N=C(C3=CC(Cl)=CC(Cl)=C3)OC2=C1)O.O[C@@H]([C@@H]([C@@H](CO)O)O)[C@@H](O)CNC
Molecular Weight (g/mol) 503.33
Formula Weight 503.33
Percent Purity 99.52%
Grade Research
Mehr anzeigen Weniger anzeigen