missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAF9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10544-1-AP-20UL
This item is not returnable.
View return policy
Description
Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals. TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. This gene encodes one of the smaller subunits of TFIID that binds to the basal transcription factor GTF2B as well as to several transcriptional activators such as p53 and VP16. A similar but distinct gene (TAF9L) has been found on the X chromosome and a pseudogene has been identified on chromosome 19. Alternative splicing results in multiple transcript variants encoding different isoforms.Specifications
| TAF9 | |
| Polyclonal | |
| Unconjugated | |
| TAF9 | |
| AD 004, AK6, CGI 137, CINAP, CIP, STAF31/32, TAF2G, TAF9, TAFII 31, TAFII 32, TAFII31, TAFII32, TAFIID32 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 108143, 373541, 6880 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q16594, Q5BKE0, Q8VI33 | |
| Taf9 | |
| TAF9 Fusion Protein Ag0833 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction