Learn More
Thermo Scientific Chemicals Tadalafil, 97%
Product Code. 16574344 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16574344 1 g 1g
16584344 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16574344 Supplier Thermo Scientific Chemicals Supplier No. 461940010
Request Bulk or Custom Format
CAS: 171596-29-5 | C22H19N3O4 | 389.41 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 171596-29-5
Molecular Formula C22H19N3O4
Molecular Weight (g/mol) 389.41
InChI Key WOXKDUGGOYFFRN-IIBYNOLFSA-N
IUPAC Name (2R,8R)-2-(2H-1,3-benzodioxol-5-yl)-6-methyl-3,6,17-triazatetracyclo[8.7.0.0³,⁸.0¹¹,¹⁶]heptadeca-1(10),11,13,15-tetraene-4,7-dione
SMILES CN1CC(=O)N2[C@H](CC3=C(NC4=CC=CC=C34)[C@H]2C2=CC=C3OCOC3=C2)C1=O
Specifications
Melting Point 296.0°C to 303.0°C
Assay Percent Range 97%
Quantity 1 g
Solubility Information Solubility aq. soln.: <1 mg/mL Other solubilities: 78 mg/mL dmso, <1 mg/mL ethanol
Formula Weight 389.4
Chemical Name or Material Tadalafil