missing translation for 'onlineSavingsMsg'
Learn More

TADA2B Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 17367-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16887244

  • 1281.92 NOK / 20µL
Estimated Shipment: 20-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

TADA2B
Polyclonal
Unconjugated
TADA2B
ADA2 beta, ADA2 like protein beta, ADA2(beta), ADA2B, TADA2B, Transcriptional adapter 2 beta
Rabbit
Antigen Affinity Chromatography
RUO
93624
-20°C
Liquid
Western Blot
0.37 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q86TJ2
TADA2B
TADA2B Fusion Protein Ag10783
20 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers