Learn More
Sampling Systems™ Tablet Counting Triangle
Product Code. 15605859 Shop All Sampling Systems Products
Change view
Click to view available options
Height (Metric):
12 mm
Unit Size:
Each
Product Code. Height (Metric) unitSize
15605859 12 mm Each
1 options
Product Code. 15605859
Brand: Sampling Systems™ A356275
Only null left
Add to basket
Add to basket
Sampling Systems™ Tablet Counting Triangle is ideal for counting tablets in hygienic and pharmaceutical applications.
  • Quick and easy way to count round tablets
  • Suitable for all sizes of round tablets
  • 316L Stainless Steel
  • Satin finish
  • Can be easily used by right or left handed operators
  • Etched with a guide to show the number of tablets in the tray
Specifications
Height (Metric) 12 mm
Width (Metric) 275 mm