Learn More
Hettich Lab Technology™ Universal 320 Benchtop Centrifuge
Description
- Versatile and durable for spinning blood and urine tubes in swing-out and angle rotors
- Up to 8 x 50 mL conical tubes for the passaging of cells in cell culture are possible
- Spins microplates for solid phase assays
- Excellent for small labs, hospitals, veterinary labs, cell culture labs, research labs and more
- Saves money and bench space
Specifications
Specifications
| Capacity | 4 x 200 mL or 6 x 94 mL |
| Max. RCF | 15,000/21,382 x g |
| Max. Speed | 15,000 rpm |
| Refrigerated | Yes |
| Model | 320 |
| Warranty | 2 Years |
| Dimensions (HWD) | 346 x 401 x 529 mm |
| Width (English) Exterior | 15.8 in. |
| Width (Metric) Exterior | 401 mm |
| Height (English) Exterior | 13.6 in. |
| Show More |